missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Graphite rod, 13.0mm (0.51 in.) dia., 99.997% (metals basis)

Brand:  Thermo Scientific Alfa Aesar 045078.DW

Graphite rod, 13.0mm (0.51 in.) dia., 99.997% (metals basis)

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15446725

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.