Learn More
SymLine™ Grounding Plug
Dovetail your SymLine™ System to suit your exact needs. Here, you will find many useful ancillaries.
Brand: SymLine™ 108281
88.30 EUR valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Additional Details : Weight : 0.01500kg
Description
SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.
- Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
- All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards
Specifications
HDPE-EC | |
GL25 (m) to 50 mm OD Pipe | |
Connection To Home Automation |
Studded | |
UK Grounding Plug (1 MΩ) with 3 x 10 mm Ports |