missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanidine hydrochloride, 99.5%, without anticaking agent, Thermo Scientific Chemicals
41.66 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Plastic Drum
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Absorbance | (6M aq. soln.), (6 M aq. soln.) at 0nm, 0.01 max. at 280nm, 0.03 max. at 260nm |
Melting Point | 178.0°C to 188.0°C |
Color | Colorless to White |
Density | 1.3450g/mL |
Infrared Spectrum | conform to structure and standard |
Assay Percent Range | 99.5% |
Packaging | Plastic bottle |
Linear Formula | H2NC(=NH)NH2·HCl |
Quantity | 100 g |
Beilstein | 03,82 |
Show More |
Safety and Handling
Product Identifier
- Guanidine hydrochloride, without anticaking agent
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'