missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Haloenol Lactone Suicide Substrate, ≥98%

For cellular and molecular biology applications

Brand:  Alfa Aesar™ J63728.MB

Code : NEW

Additional Details : CAS Number : 88070-98-8 Weight : 0.00025kg

 View more versions of this product

Product Code. 15415919

  • 675.00 € / 25mg


Estimated Shipment date
from Supplier 30-07-2018
Add to basket

Specifications

Specifications

White
Lyophilized
25mg
Irreversibly inhibits calcium-independent phospholipase A-2
317.18
-30° to -10°C
Videos
SDS

For Research Use Only