Learn More
Hamilton™ Microlab™ Accessories - PTFE Tubing Assemblies
For use with Hamilton Microlab 600 Diluters/Dispenser or Microlab 300 Pipetting System
42.00 € - 98.00 €
Specifications
Material | FEP |
---|---|
For Use With (Equipment) | Microlab™ 600 diluter/dispenser system |
Includes | 1/4-28 Tubing threads |
Product Code | Brand | Volume (Metric) | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Type | Price | Quantity & Availability | ||||
12658333
|
Hamilton™
61614-01 |
4.57 mL | Gauge 12, Fill Tubing |
42.00 €
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
12346249
|
Hamilton™
61615-01 |
1.15 mL | Gauge 18, Fill Tubing |
44.00 €
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
12698333
|
Hamilton™
240133 |
5.15 mL | Gauge 12, Dispense Tubing |
48.00 €
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
11318421
|
Hamilton™
240134 |
1.29 mL | Gauge 18, Dispense Tubing |
52.00 €
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
15339712
|
Hamilton™
1174-02 |
0.94 μL/mm | Gauge 18, Fill Tubing |
60.00 €
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
12398809
|
Hamilton™
1173-02 |
0.94 μL/mm | Gauge 18, Dispense Tubing |
74.00 €
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
15329712
|
Hamilton™
1171-02 |
3.75 μL/mm | Gauge 12, Dispense Tubing |
74.00 €
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
15399712
|
Hamilton™
61491-01 |
3.75 μL/mm | Gauge 12, Continuous Fill Tubing |
98.00 €
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
15319722
|
Hamilton™
61491-02 |
0.94 μL/mm | Gauge 18, Continuous Fill Tubing |
88.00 €
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
||||
Description
- For use with Hamilton Microlab 600 Diluters/Dispenser or Microlab 300 Pipetting System
Specifications
FEP | |
Microlab™ 600 diluter/dispenser system | |
1/4-28 Tubing threads |