Learn More
Hamilton™ Microlab™ Accessories - PTFE Tubing Assemblies
For use with Hamilton Microlab 600 Diluters/Dispenser or Microlab 300 Pipetting System
Brand:  Hamilton™ 61491-02
Code : NEW
Additional Details : Weight : 0.01600kg
 View more versions of this product
Product Code. 15319722
  • 88.00 € / Each
Estimated Shipment: 28-05-2024
to see stock.
Add to basket
Description
Description
  • For use with Hamilton Microlab 600 Diluters/Dispenser or Microlab 300 Pipetting System
Specifications
Specifications
FEP
0.94 μL/mm
1/4-28 Tubing threads
Gauge 18, Continuous Fill Tubing
Microlab™ 600 diluter/dispenser system