Learn More
Bellingham Stanley™ Hand Held Refractometer
Premium quality instruments suitable for use in the harshest of environments
POA
Specifications
Quantity 1 Each
View More Specs
Description
Description
OPTi hand held, digital refractometers are highly reliable, premium quality instruments suitable for use in the harshest of environments. Constructed using the latest manufacturing techniques including stainless steel injection moulding to construct the easy clean prism dish, ultrasonic welding to bond the housings and a rubberised switch membrane to further protect against moisture ingress and excess wear. May be used to control the dissolved solids or blend ratios of numerous products from fresh fruit to industrial chemicals.
  • Single and duo scale models
  • Zero calibration with water
  • Shallow prism dish
  • Ergonomic design with small hand print
For analysing food, beverages, fruit and chemicals, also has applications in the automotive industry.
Specifications
Specifications
1 Each