missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Hardboard cover for tubular shelving, 1000 x 400mm
Brand: Manutan A360400
Code : Z3
Additional Details : Weight : 1.00000kg
Description
Ideal for the storage of smaller items, 3mm thick. Use to cover tubular shelves with easy click-in system.- Thickness: 3mm
- Depth: 400mm