Learn More
Brady™ Hazardous Chemical Labels for B30 Series
Pre-printed, GHS/CLP chemical labels for BBP3X, S3XXX, and i3300 printers.
Brand:  Brady™ 121649
Additional Details : Weight : 1.34490kg
 View more versions of this product
Product Code. 15931011
  • 425.00 € / Pack of 200
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Description
Description
  • Permanent acrylic adhesive
  • For smooth and textured surfaces
Specifications
Specifications
Not Autoclavable
Red on White
Gloss
Thermal Transfer
Facility Identification, Hazard Communication Identification, Electrical Labeling
1
Rectangle
0.102 mm
155 mm
Permanent Acrylic
L4A Layout
Vinyl (B-7569)
Non-sterile
BBP™30, BBP™31, BBP™33, BBP™35 Multicolor, BBP™37 Multicolor, BradyPrinter i3300, BradyPrinter S3000, S3100, and i5300 Industrial Label Printers
200 Labels
-20°C to +75°C
162 mm
105 mm