missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Key Industrial Equipment™ Hazardous storage cabinet, for acid/alkali, white, 1830mm x 915mm x 459mm
Brand: Key Industrial Equipment™ A350585
Code : Z3
Additional Details : Weight : 1.00000kg
Description
Hard wearing cabinets suitable for storing mild acids and corrosive chemicals. Finished in white polyester powder coating, resistant to most acids and alkalis.- Three shelves
- Dimensions (H x W x D) : 1830mm x 915mm x 459mm