missing translation for 'onlineSavingsMsg'
Learn More

PEPperCHIP™ HCMV Antigen Microarray

HCMV Antigen Microarray with 4327 antigen specific peptides printed in duplicate. The microarray can be used for epitope mapping, serum screening or biomarker discovery.

Brand:  PEPperCHIP™ SPC.012.001

Additional Details : Weight : 0.02200kg

Product Code. 16347758

  • 560.74 € / Each
Estimated Shipment: 12-07-2024
to see stock.
Add to basket

Description

Description

  • Microarray Content: 24 selected antigens of human cytomegalovirus (HCMV): 28kDa structural phosphoprotein, major capsid protein, protein UL40, 150kDa matrix phosphoprotein, Orf UL153, 65kDa matrix phosphoprotein, protein UL122, 55kDa immediate-early protein 1, protein UL128, protein UL130, UL131A, Envelope glycoprotein B, protein UL36, protein UL28, large tegument protein, tegument protein UL94, protein UL103, glycoprotein E, protein HHLF6, protein HHRF4, protein HHRF7, Orf UL151
  • Peptide Length/Overlap: 15 aa/ 12 aa
  • Number of Peptides: 4327 printed in duplicate and HA, polio and c-Myc control peptides
Specifications

Specifications

PEPperSlide™ PEGMA/PMMA
Antibody validation, epitope mapping, serum screening, biomarker discovery
Microscopic Glass
Individual barcode for each microarray
1 Slide
1 mm
Transparent
75.4 x 25 mm
High-density Peptide Microarray
Non-sterile
1 array copy per microarray
Combinatorial peptide synthesis by amino acid laser printing
ISO 9001:2015
RUO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers