missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ HDPE Wide Neck Square Reagent Bottles

Product Code. 11827702 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
750 mL
1000 mL
1500 mL
2500 mL
4000 mL
Unit Size:
Each
Pack of 6
This item is not returnable. View return policy

Product Code. 11827702

Brand: Buerkle™ 03421000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The Wide-neck reagent bottle is particularly suited for storing, packing, taking samples, sampling and transport of granules and paste-like media.

  • HDPE, transparent
  • PP tamper-evident screw cap, blue
  • UN approval for bulk goods
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1000 mL
Color Transparent
Diameter (Metric) Inner Neck 43 mm
Material HDPE
Length (Metric) 85 mm
Width (Metric) 85 mm
Thread Size (Metric) 54 mm
Closure Color Blue
Certifications/Compliance UN Approval
Closure Included Yes
Mouth Wide
Height (Metric) 214 mm
Closure Material Polypropylene
Closure Type Screw Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.