missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ HDPE Wide Neck Square Reagent Bottles
Shop All Bürkle Products
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
750 mL
1000 mL
1500 mL
2500 mL
4000 mL
Unit Size:
Each
Pack of 6
Description
- HDPE, transparent
- PP tamper-evident screw cap, blue
- UN approval for bulk goods
Specifications
Specifications
| Capacity (Metric) | 100 mL |
| Color | Transparent |
| Diameter (Metric) Inner Neck | 24 mm |
| Material | HDPE |
| Length (Metric) | 45 mm |
| Width (Metric) | 45 mm |
| Thread Size (Metric) | 32 mm |
| Closure Color | Blue |
| Certifications/Compliance | UN3H1/x0.3 |
| Closure Included | Yes |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction