missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Heavy duty roller bearing shutter security cabinet, green, louvred, 2000mm x 1000mm x 500mm
Brand: Manutan 381B197
Code : Z3
Additional Details : Weight : 1.00000kg
Description
Built to endure tough working environments. Heavy duty welded construction in 1. 2mm thick sheet steel. Counter balanced galvanised steel roller bearing shutter door assembly. Secured by matching heavy duty security pin locks.- Green door
- Dimensions (H x W x D): 2000mm x 1000mm x 500mm