missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Hei-VAP Industrial Rotary Evaporator with Glassware Set

Hei-VAP Industrial rotary evaporators have excellent user safety features and are available with a variety of glassware sets.

Brand:  Heidolph Instruments 518-18100-00


Code : A0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10027851

  • POA / Each
Estimated Shipment: 11-05-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

  • Excellent user safety with high-impact transparent PMMA door
  • Non-fogging safety glass and metal frame guard hood provides excellent user protection
  • The evaporation flask is illuminated during operation for increased visibility
  • Universal heating bath accommodates water or other bath fluids allowing for temperature settings up to 180°C
  • Receiver cassettes and additional PMMA door housing protect against threat of glassware breakage
  • Comes as standard with integrated refill water system, spillover prevention and a release valve at the bottom
  • Optional Base Cart is fully-maneuverable with removable panels for storage of vacuum pump, glassware, solvents or accessories
  • Certification according to GMP available for this model: validation for installation (IQ) and operating qualification (OQ)
  • Large touch screen control panel with illuminated displays for all process parameters, programmable ramps
Specifications

Specifications

6 to 160 rpm
7 in. touchpanel
Glassware set Shorty R - (1) ascending condenser (320 mm shorter than traditional one), (1) 20 L evaporating flask, (1) 10 L receiving flask
4000 W
20°C to 180°C
Ideal in laboratories for hood applications or where height is restricted
IP20
Glassware Assembly Rotary Evaporator
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers