Learn More
Hirschmann™ Sodium Heparinized Hematocrit Tubes
Excellent for centrifuging blood in the laboratory. Hirschmann Instruments™ Sodium Heparinized Hematocrit Tubes are manufactured from disposable capillary glass.
Brand: Hirschmann™ 9100299
Additional Details : Weight : 0.80000kg
Description
- Color-coded red
- Acc. to DIN EN ISO 12772, CE acc. IVD 98/79 EC
Specifications
Hematocrit Tube Na-hep | |
100 μL | |
For centrifuging blood samples, Sodium heparinized | |
DIN EN ISO 12772, CE, IVD 98/79 EC |