Learn More
Hirschmann™ Sodium Heparinized Hematocrit Tubes
Excellent for centrifuging blood in the laboratory. Hirschmann Instruments™ Sodium Heparinized Hematocrit Tubes are manufactured from disposable capillary glass.
Brand: Hirschmann™ 9100260
Additional Details : Weight : 0.26000kg
Description
- Color-coded red
- Acc. to DIN EN ISO 12772, CE acc. IVD 98/79 EC
Specifications
Hematocrit Tube Na-hep | |
0.9/1.0 mm | |
75 ±0.5 mm |
60 μL | |
For centrifuging blood samples, Sodium heparinized | |
DIN EN ISO 12772, CE, IVD 98/79 EC |