missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Hematoxylin hydrate, 80% (dry wt.), water <8%

Brand:  Alfa Aesar™ 047223.22

Code : NEW

Additional Details : CAS Number : 517-28-2 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 15472917

  • 289.00 € / 100g


Estimated Shipment date
from Supplier 22-05-2019
Add to basket

Specifications

Chemical Identifiers

517-28-2
302.282
WZUVPPKBWHMQCE-VYIIXAMBSA-N
45029742
C1C2=CC(=C(C=C2C3C1(COC4=C3C=CC(=C4O)O)O)O)O
C16H14O6
MFCD00078111
hematoxylin, hydroxybrasilin
(6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol

Specifications

Hematoxylin hydrate
Brown to Cream
100g
517-28-2
C16H14O6
91399
hematoxylin, hydroxybrasilin
C1C2=CC(=C(C=C2C3C1(COC4=C3C=CC(=C4O)O)O)O)O
302.282
302.29 (Anhydrous)
80%
Light Sensitive
~200°C (decomposition)
Water <8%
(dry weight)
MFCD00078111
14,4637
WZUVPPKBWHMQCE-VYIIXAMBSA-N
(6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol
45029742
Powder
Videos
SDS