missing translation for 'onlineSavingsMsg'
Learn More

Hematoxylin hydrate, 80% (dry wt.), water <8%, Thermo Scientific Chemicals

Catalog Number p-7052339
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
517-28-2
C16H14O6
302.28
MFCD00078111
WZUVPPKBWHMQCE-WKTCHCBJNA-N
hematoxylin, hydroxybrasilin
45029742
(6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol
[H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3
This item is not returnable. View return policy
517-28-2
C16H14O6
302.28
MFCD00078111
WZUVPPKBWHMQCE-WKTCHCBJNA-N
hematoxylin, hydroxybrasilin
45029742
(6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol
[H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3

Hematoxylin and eosin together make up Hematoxylin and eosin stain, one of the most commonly used stains in histology. Metal-haematein complexes are used to stain cell nuclei prior to examination under a microscope. Structures that stain with iron- or aluminum-haematein are often called basophilic, even though the mechanism of the staining is different from that of staining with basic dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼200°C (decomposition)
Color Brown to Cream
Quantity 5 g
Beilstein 91399
Sensitivity Light sensitive
Merck Index 14,4637
Solubility Information Soluble in 95% ethanol(1mg/mL).
Formula Weight 302.29 (Anhydrous)
Percent Purity 80%
Physical Form Powder
Assay (dry weight)
Chemical Name or Material Hematoxylin hydrate, Water <8%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'