missing translation for 'onlineSavingsMsg'
Learn More

Hematoxylin Trihydrate, Fisher BioReagents

Catalog Number 10012694
Click to view available options
Quantity:
5 g
100 g
Packaging:
Amber Glass
This item is not returnable. View return policy
517-28-2
C16H14O6
302.28
MFCD00078111
WZUVPPKBWHMQCE-WKTCHCBJNA-N
Natural Black I
45029742
[H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3
This item is not returnable. View return policy
517-28-2
C16H14O6
302.28
MFCD00078111
WZUVPPKBWHMQCE-WKTCHCBJNA-N
Natural Black I
45029742
[H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3
Hematoxylin Trihydrate is used mainly as a stain in microscopy.
TRUSTED_SUSTAINABILITY
Melting Point 100°C
Merck Index 15, 4672
Identification Pass Test
IUPAC Name (1R,10S)-8-oxatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-2(7),3,5,12(17),13,15-hexaene-5,6,10,14,15-pentol
Lambda Max 290 to 292nm
Formula Weight 356.4
Loss on Drying Report %
Packaging Amber Glass
Color Amber
Quantity 5 g
Physical Form Powder
Chemical Name or Material Hematoxylin Trihydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'