Learn More
Hemokinin 1, Human, Thermo Scientific™
For research use
Brand: Thermo Scientific Alfa Aesar J65288.#0
Code : NEW
Additional Details : CAS Number : 491851-53-7 Weight : 0.00005kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Lyophilized powder | |
-30° to -10°C | |
Hemokinin 1, Human | |
Lyophilized |
5mg | |
RUO | |
Native |
For Research Use Only