missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HEPA Filter Kit

For Use With: Steri-Cult 3307/3308, 3310/3311 Incubators

Brand:  Thermo Scientific™ LT1900160

Code : 50

Additional Details : Weight : 2.16000kg

Product Code. 10748693

  • 371.00 € / Each
In Stock
to see stock.
Add to basket

Description

Description

Help ensure measurable Class 100 air quality performance with Thermo Scientific™ Replacement HEPA Filters and Cleaning Kits.

HEPA Filters

  • 99.97% filtering at 0.3μm
  • Compatible with Thermo Scientific™ Forma™ Steri-Cult™ 3307 and 3310 CO2 Incubators
  • Main chamber filter available as a stand-alone or packaged in a group with the other chamber in-line filters

Decontamination Kits

  • Consists of filters for difficult-to-clean chambers and gaskets (except door) and tubing
  • Reduces downtime and labor spent on decontamination
Specifications

Specifications

HEPA Filter Replacement Kit
Steri-Cult™ 3307 and 3310 CO2 Incubators
LED
HEPA Filter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers