Learn More
Thermo Scientific™ Heracell™ Split Shelf System
Material: Copper; For Use With: Heracell™ VIOS CO2 Incubators
Brand:  Thermo Scientific™ 51901122
Additional Details : Weight : 0.10000kg
Product Code. 15542720
  • 734.00 € / Each
Estimated Shipment: 12-08-2024
to see stock.
Add to basket
Description
Description
Half shelf system, Split Shelf; 6 half-width in place of 3 full -width, Material: copper; For use with Heracell VIOS 250i CO2 Incubator
  • For use with 6 gas tight inner door configuration
  • Half shelf system
  • 6 half-width in place of 3 full-width
Specifications
Specifications
6 split copper shelves
LED
Accessory, Shelf System
Heracell™ VIOS CO2 Incubators
Copper