missing translation for 'onlineSavingsMsg'
Learn More

Hexafluoroglutaric anhydride, 97%, Thermo Scientific Chemicals

Catalog Number 11337845
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
376-68-1
C5F6O3
222.042
MFCD00006680
IHYAGCYJVNHXCT-UHFFFAOYSA-N
hexafluoroglutaric anhydride, perfluoroglutaric anhydride, 3,3,4,4,5,5-hexafluorodihydro-2h-pyran-2,6 3h-dione, hexafluoroglutaric acid anhydride, 2,2,3,3,4,4-hexafluoropentanedioic anhydride, 3,3,4,4,5,5-hexafluoro-3h-4,5-dihydropyran-2,6-dione, hexafluoroglutaricanhydride, acmc-1clz4, hexafluorooxane-2,6-dione
78995
3,3,4,4,5,5-hexafluorooxane-2,6-dione
C1(=O)C(C(C(C(=O)O1)(F)F)(F)F)(F)F
This item is not returnable. View return policy
376-68-1
C5F6O3
222.042
MFCD00006680
IHYAGCYJVNHXCT-UHFFFAOYSA-N
hexafluoroglutaric anhydride, perfluoroglutaric anhydride, 3,3,4,4,5,5-hexafluorodihydro-2h-pyran-2,6 3h-dione, hexafluoroglutaric acid anhydride, 2,2,3,3,4,4-hexafluoropentanedioic anhydride, 3,3,4,4,5,5-hexafluoro-3h-4,5-dihydropyran-2,6-dione, hexafluoroglutaricanhydride, acmc-1clz4, hexafluorooxane-2,6-dione
78995
3,3,4,4,5,5-hexafluorooxane-2,6-dione
C1(=O)C(C(C(C(=O)O1)(F)F)(F)F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.652
Boiling Point 71°C to 72°C
Flash Point None
Odor Pungent
Refractive Index 1.324
Quantity 1 g
UN Number UN3265
Beilstein 210699
Sensitivity Moisture sensitive
Formula Weight 222.04
Percent Purity 97%
Chemical Name or Material Hexafluoroglutaric anhydride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'