missing translation for 'onlineSavingsMsg'
Learn More

Hexafluorophosphoric acid, ca 60% w/w aq. soln.

Product Code. 11392757
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
This item is not returnable. View return policy

Product Code. 11392757

Brand: Thermo Scientific Alfa Aesar L15728.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16940-81-1 | HPF6

Hexafluorophosphoric acid is used as a catalyst. It is also used as a metal cleaner and chemical polishing agent for the formation of protective coatings of metal surfaces..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexafluorophosphoric acid is used as a catalyst. It is also used as a metal cleaner and chemical polishing agent for the formation of protective coatings of metal surfaces..

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with strong bases, glass, alkali metals, powdered metals, strong oxidizing agents, metals and reacts violently with water.  
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16940-81-1
Molecular Formula HPF6
MDL Number MFCD00011350

Specifications

Density 1.65
Quantity 500 mL
UN Number UN1782
Solubility Information Immiscible with water.
Formula Weight 145.97
Concentration or Composition (by Analyte or Components) ≈60% w/w aq. soln.
Chemical Name or Material Hexafluorophosphoric acid
compliance-icons compliance-icons
Product Identifier
  • Hexafluorophosphoric acid, ca 60% aqueous solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P262-Do not get in eyes, on skin, or on clothing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Hexafluorophosphoricacid, Hydrofluoricacid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.