Learn More
Hexamethyldisilazane, Electronic grade, 99+%, Thermo Scientific™
69.40 € - 189.00 €
Chemical Identifiers
| CAS | 999-97-3 |
|---|---|
| Molecular Formula | C6H19NSi2 |
| Molecular Weight (g/mol) | 161.395 |
| MDL Number | MFCD00008259 |
| InChI Key | FFUAGWLWBBFQJT-UHFFFAOYSA-N |
| Synonym | hexamethyldisilazane, bis trimethylsilyl amine, hmds, 1,1,1,3,3,3-hexamethyldisilazane, hexamethylsilazane, silanamine, 1,1,1-trimethyl-n-trimethylsilyl, tri-sil, 1,1,1-trimethyl-n-trimethylsilyl silanamine, hexamethyldisilizane, disilazane, 1,1,1,3,3,3-hexamethyl |
| PubChem CID | 13838 |
| ChEBI | CHEBI:85068 |
| IUPAC Name | [dimethyl-(trimethylsilylamino)silyl]methane |
| SMILES | C[Si](C)(C)N[Si](C)(C)C |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
11389697
|
Thermo Scientific Alfa Aesar
042039.22 |
100g |
69.40 €
100g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
11309707
|
Thermo Scientific Alfa Aesar
042039.36 |
500g |
189.00 €
500g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identificadores de productos químicos
| C6H19NSi2 | |
| MFCD00008259 | |
| hexamethyldisilazane, bis trimethylsilyl amine, hmds, 1,1,1,3,3,3-hexamethyldisilazane, hexamethylsilazane, silanamine, 1,1,1-trimethyl-n-trimethylsilyl, tri-sil, 1,1,1-trimethyl-n-trimethylsilyl silanamine, hexamethyldisilizane, disilazane, 1,1,1,3,3,3-hexamethyl | |
| CHEBI:85068 | |
| C[Si](C)(C)N[Si](C)(C)C |
Especificaciones
| 999-97-3 | |
| 126°C | |
| Odorless | |
| (CH3)3SiNHSi(CH3)3 | |
| MFCD00008259 | |
| 635752 | |
| 14,4689 | |
| Miscible with acetone,benzene,ethyl ether,heptane and perchloroethylene. | |
| C[Si](C)(C)N[Si](C)(C)C | |
| 161.395 | |
| CHEBI:85068 | |
| ≥99% | |
| Hexamethyldisilazane |
| 0.774 g/mL | |
| 8°C (46°F) | |
| C6H19NSi2 | |
| 1.408 | |
| UN3286 | |
| Moisture sensitive | |
| hexamethyldisilazane, bis trimethylsilyl amine, hmds, 1,1,1,3,3,3-hexamethyldisilazane, hexamethylsilazane, silanamine, 1,1,1-trimethyl-n-trimethylsilyl, tri-sil, 1,1,1-trimethyl-n-trimethylsilyl silanamine, hexamethyldisilizane, disilazane, 1,1,1,3,3,3-hexamethyl | |
| FFUAGWLWBBFQJT-UHFFFAOYSA-N | |
| [dimethyl-(trimethylsilylamino)silyl]methane | |
| 13838 | |
| 161.4 | |
| Electronic |
Seguridad y manipulación
GHS H Statement
H225-H311-H331-H314-H318-H302
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302+H332-H311-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 213-668-5
missing translation for 'rtecsNumber' : JM9230000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.