missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylenetetramine, 98.5%, contains an anticaking agent, Thermo Scientific Chemicals

Catalog Number p-142065
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
100-97-0
C6H12N4
140.19
MFCD00006895
VKYKSIONXSXAKP-UHFFFAOYSA-N
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine
4101
CHEBI:6824
C1N2CN3CN1CN(C2)C3
This item is not returnable. View return policy
100-97-0
C6H12N4
140.19
MFCD00006895
VKYKSIONXSXAKP-UHFFFAOYSA-N
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine
4101
CHEBI:6824
C1N2CN3CN1CN(C2)C3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 260.0°C
Color White
Density 1.3300g/mL
Flash Point 250°C
Infrared Spectrum Authentic
Assay Percent Range 98.5%
Packaging Plastic bottle
Quantity 1 kg
Beilstein 01, 583
Fieser 01,427; 02,208; 04,243; 09,234
Merck Index 15, 6038
Specific Gravity 1.33
Solubility Information Solubility in water: 895g/L (20°C). Other solubilities: soluble in chloroform: 13.4% (20°C), soluble in ethanol: 2.89% (20°C), soluble in acetone: 0.65% (20°C), insoluble in ether: 0.06% (20°C), soluble in methanol: 7.25% (20°C)
Sublimation Point 263.0 °C
Formula Weight 140.19
Percent Purity 98.5%
Physical Form Crystalline Powder
Chemical Name or Material Hexamethylenetetramine, 98.5%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'