Learn More
Gilson™ HF2 Pump Head for Gilson™ Peristaltic Pump
Product Code. 12316122 Shop All Gilson Products
Click to view available options
Flow Rate:
1 to 340 mL/min.
Unit Size:
Each
Product Code. 12316122
Brand: Gilson™ F110707
Only null left
Add to basket
Add to basket
Add channels to your MINIPULS™ 3 or Evolution and allow even greater flexibility in your laboratory.
MINIPULS 3 Pump Heads
Choose from these MINIPULS 3 pump heads:
  • R1—Single channel pump head
  • R4—Four channel pump head
  • R8—Eight channel pump head
  • R2—Two channel pump head
  • R2HF—Two channel high flow pump head
  • R4HF—Four channel high flow pump head
MINIPULS Evolution Pump Heads
Choose from these MINIPULS Evolution pump heads:
  • MF1—Pump head with one cartridge
  • MF4—Pump head with four cartridges
  • HF2—Pump head with two high flow cartridges
TRUSTED_SUSTAINABILITY
Specifications
Flow Rate 1 to 340 mL/min.