missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Hi-Temp Vacuum Ovens
Ideal for higher temperature drying under vacuum, this vacuum ovens line provides temperatures up to 280°C, with peace of mind due to built-in safety controller and overtemperature protection.
POA
Specifications
Chamber Material | Stainless Steel |
---|---|
Type | Vacuum Oven |
Display | LED (2) |
Material | Powder-coated Heavy-gauge Steel |
No. of Shelves | 2 |
Description
Ideal for higher temperature drying under vacuum, this vacuum ovens line provides temperatures up to 280°C, with peace of mind due to built-in safety controller and overtemperature protection.
Features include:- Two convenient sizes: Small and large model, 18L (0.65 cu.ft.) and 42.4L (1.5 cu.ft.)
- Temperature range: Ambient +10°C / 50°C to 280°C
- Radiant warm wall heaters mounted on the outside conserve chamber working space and help eliminate potential hazards of open wire heaters
- Compression fittings on the side of the unit accept 6.4mm (0.25 in.) outer diameter hard tubing for pump and air connections
- Vacuum and air lines of corrosion-resistant stainless-steel tubing optimize chamber cleanliness and long-term performance
- Durable exterior is powder-coated heavy-gauge steel
- Glass window in the door allows for sample viewing
- Two adjustable chrome-plated shelves help improve heat conduction or transfer to samples; easily removable for cleaning
Small models 3625A and 3625A-1
- Dual-electronic control system with dial thermometer (no display)
- For operational convenience, all controls and indicators are located on the front panel: power switch and three indicators, purge gas and vacuum control valves, a 0-to-30-inch vacuum gauge and a temperature control
Large models 3628A and 3628A-1
- Single set point controller
- Seven-day programmable timer that cycles oven on or off up to eight times per day with automatic temperature/ vacuum control in real time steps
- Dual displays provide constant day/ time and temperature/pressure updates every 5 seconds
Recommended for:
- Desiccation
- Vacuum embedding
- Plating
- Process control
- Inert atmosphere
Specifications
Stainless Steel | |
LED (2) | |
2 | |
50/60 Hz |
Vacuum Oven | |
Powder-coated Heavy-gauge Steel | |
Chrome-plated |