Learn More
Emma Safety Footwear Himalaya XD Safety Shoes
A modern shoe that not only looks good but also protects your feet from top to bottom and everywhere else. Himalaya features a steel toe cap and an integrated stainless steel plate. The double-layer, anti-slip PU sole provides extra protection from hazards on the ground.
Brand:  Emma Safety Footwear MM33556844
Additional Details : Weight : 0.40000kg
 View more versions of this product
Product Code. 15905150
  • 133.00 € / 1 pair
Estimated Shipment: 26-06-2024
to see stock.
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Oil Nubuck construction
  • GripForce™ Easy Twist
  • 3M™ Reflective Striping
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Specifications
Himalaya XD
Oiled Nubuck
44