missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiScale™ Column Tubes
608.00 € - 1542.00 €
Specifications
Material | Borosilicate Glass, Polycarbonate, Polyamide (Reinforced Glass Fiber), Ethylene Propylene Diene Poly |
---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
12415690
|
Cytiva
28-9666-51 |
26 mm |
1002.00 €
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
12425690
|
Cytiva
28-9666-52 |
16 mm |
608.00 €
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
12405690
|
Cytiva
28-9666-50 |
50 mm |
1542.00 €
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Specifications
Borosilicate Glass, Polycarbonate, Polyamide (Reinforced Glass Fiber), Ethylene Propylene Diene Poly |