missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap Capto S ImpAct Prepacked Columns

HiTrap Capto S ImpAct columns are prepacked with Capto S ImpAct cation exchange chromatography medium for screening and small-scale protein purifications using ion exchange chromatography (IEX).

Brand:  Cytiva 17371755

Code : NEW

Additional Details : CAS Number : 64-17-5 Weight : 0.12200kg


 View more versions of this product

Product Code. 15371642

  • 929.00 € / Pack of 5
Estimated Shipment: 18-06-2024
to see stock.
Add to basket

Description

Description

  • Convenient prepacked column for fast, reproducible, and easy small-scale purifications.
  • High binding capacity, > 100 mg MAb/mL medium.
  • Efficient aggregate removal at high load of monoclonal antibodies.
  • High-resolution intermediate purification and polishing based on the well-established Capto platform.
  • Flexibility of design thanks to a large operational window of flow rates and bed heights for easy optimization and scaling.
Specifications

Specifications

HiTrap Capto S ImpAct Prepacked Column
20mL/min
4 to 12
Ion Exchange Column
5 x 5mL
High-flow agarose
50μm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiTrap Capto S ImpAct, 5 ml, 5 x 5 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-18 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers