Learn More
Cytiva HiTrap IXSelect
HiTrap IXSelect columns are prepacked with IXSelect resin for screening and small-scale protein purifications using affinity chromatography.
Brand:  Cytiva 17371411
Additional Details : Weight : 0.08000kg
 View more versions of this product
Product Code. 15579261
  • 1022.00 € / Pack of 5
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
Description
Description
  • XSelect is a high-throughput resin specifically designed for the purification of factor IX targets from recombinant feed streams.
  • Columns for screening of selectivity, binding and elution conditions, as well as small scale purifications.
Specifications
Specifications
2°C to 8°C
5 x 1 mL