Learn More
HNHA, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J64917.MA
Code : NEW
Additional Details : CAS Number : 926908-04-5 Weight : 0.00010kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
A cell-permeable hydroxyamide-based SAHA analog that potently inhibits histone deacetylase (HDAC) activity.Specifications
10 mg | |
A cell-permeable hydroxyamide-based SAHA analog that potently inhibits histone deacetylase (HDAC) activity | |
C17H21NO2S | |
Histone Deacetylase Inhibitor VI | |
Signal Transduction Reagent |
2°C to 8°C | |
926908-04-5 | |
303.42g/mol | |
Light Sensitive | |
Crystalline Solid |