missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units

Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems

Brand:  Cytiva SE511-12-1.5

Code : 04

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11955348

  • 81.08 € / Each


Estimated Shipment
03-06-2021
Add to basket

Description

Description

  • Lightweight
  • Virtually unbreakable

  • Come attached to a standard back which produces maximum well depths of 25mm, except for the 28-well preparative combs which produce well depths of 15mm
  • Adjustable comb back (available separately, below) converts 25mm deep combs to 10 or 15mm depth

Specifications

Specifications

0.98 in.
SE 600, SE 400 Series Vertical Electrophoresis System
PTFE
0.05 in.
11.5μL/ mm
2.5 cm
Comb
12
0.15 cm
0.76 cm
Videos
SDS
Documents