missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ Glass Plates for SE 600 Chroma, SE 600 Standard, or SE 400 Vertical Electrophoresis Units
Accessory parts for SE 600 Chroma SE 600 Standard or SE 400 Vertical Electrophoresis Units
65.25 € - 160.00 €
Specifications
For Use With (Equipment) | SE 600 Chroma, SE 600 Standard, SE 400 Vertical Electrophoresis Unit |
---|---|
Material | Glass |
Quantity | 2/Pk. |
Dimensions (L x W) | 18 x 16 cm |
Length (Metric) | 18 cm |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
11519753
|
Hoefer™
SE6102 |
Glass Plate |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11905368
|
Hoefer™
SE6102LF |
Low Fluorescence Glass Plate |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- For use with SE660, SE600X Chroma, SE600, SE640, SE410, and SE400 Vertical Electrophoresis Units
- Regular (straight, without notch)
- Supplied in packs of 2
Specifications
SE 600 Chroma, SE 600 Standard, SE 400 Vertical Electrophoresis Unit | |
2/Pk. | |
18 cm |
Glass | |
18 x 16 cm | |
16 cm |