Learn More
Hoefer™ Mighty Small™ II Mini Vertical Electrophoresis Systems
Combine the advantages of a small format and efficient cooling for rapid screening of protein and nucleic acids
Brand: Hoefer™ SE250-10A-1.0
Code : 04
Additional Details : Weight : 0.54000kg
Includes: SE250 Basic System includes: upper buffer chamber, lower buffer chamber, interlocking safety lid with leads, ten 10 x 8cm glass plates, two 10 x 8cm notched alumina plates, four spring clamps, gel seal, well locating decal, and Spacer Mate. SE250 Complete System also includes: Dual Gel Caster, two 0.75mm thick 10-well combs, and two pairs 0.75mm thick spacers. SE260 Basic System includes same items as SE250 Basic Systems, except the glass and alumina plates.SE260 Complete Systems add Dual Gel Caster, ten 10.5 x 10cm glass plates, five 10.5 x 10cm notched alumina plates, and two 10-well combs and two pairs spacers (either 0.75mm or 1.5mm thick; see ordering table).
Description
Combine the advantages of a small format and efficient cooling for rapid screening of protein and nucleic acids
- Run two gels at once
- Produce high-resolution separations in as little as 45 minutes with no band “smiling”
- Use alumina plates (included with specified models) for efficient cooling
- Buffer volume: 300 mL
Applications:
Ideal for uniform and gradient acrylamide, agarose, IEF, field inversion agarose and 2-D electrophoresis
Certifications
CE Marked, CSA C22.2 1010.1, EN61010-1, UL3101-1.
Specifications
EN61010-1, UL61010A-1, CSA C22.2 1010.1, CE | |
Up to 80% | |
8 cm | |
18 cm | |
12 w |
Electrophoresis Unit | |
Mini Vertical Electrophoresis Unit, complete with 1mm Accessories | |
7 in. | |
500 V |