Learn More
Honeywell™ XC™ Clear FogBan™ Safety Glasses
Catalog Number 11816940
Click to view available options
Blue Frames
Provides total wraparound coverage and ensures exceptional peripheral vision. Sperian™ Honeywell™ XC™ Clear FogBan™ Safety Glasses protect from the top of the eye to the bottom with no gaps.
  • Adjustable temples and lenses with flexible nose piece
  • Extra flexible frame of blue plastic
  • Easily replaceable lenses - can be changed without replacing the entire frame
  • All pressure points are relieved for exceptional all-day comfort
TRUSTED_SUSTAINABILITY
Certifications/Compliance EN 166 and EN 170 1F 3-1.2
Lens Coating Anti-fog
Lens Tint Clear
Product Type Eye and Face Protection Spectacles
For Use With (Application) General use and protection against impact (45m/s), Mechanical work (grinding, turning, milling)
Frame Color Blue
No. per Pack 1 Ea.
Lens Material Polycarbonate
Frame Material Plastic