Learn More
Epredia™ HP35n Ultra High-profile Disposable Microtome Blades
Epredia™ HP35n ultra high-profile disposable microtome blades are good for routine tissues when the user is used to working with non-coated blades.
Brand: Epredia™ 3151735
Code : NEW
Additional Details : Weight : 0.50000kg
Description
- Ultra High-profile
- Non-coated
- Highest Durability
Specifications
High Profile Blade | |
Cryostat | |
None |
High | |
Yes | |
35° |