Learn More
Abnova™ Human ADCYAP1 Partial ORF (NP_001108.1, 95 a.a. - 172 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_001108.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 116 |
Molecular Weight (g/mol) | 34.32kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16043595
|
Abnova™
H00000116-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16033595
|
Abnova™
H00000116-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes adenylate cyclase activating polypeptide 1. Mediated by adenylate cyclase activating polypeptide 1 receptors, this polypeptide stimulates adenylate cyclase and subsequently increases the cAMP level in target cells. Adenylate cyclase activating polypeptide 1 is not only a hypophysiotropic hormone, but also functions as a neurotransmitter and neuromodulator. In addition, it plays a role in paracrine and autocrine regulation of certain types of cells. This gene encodes three different mature peptides, including two isotypes, a shorter form and a longer form. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]
Sequence: VLDQLSAGKHLQSLVARGVGGSLGGGAGDDAEPLSKRHSDGIFTDSYSRYRKQMAVKKYLAAVLGKRYKQRVKNKGRRSpecifications
NP_001108.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.32kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC126852/PACAP | |
ADCYAP1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
116 | |
ADCYAP1 (Human) Recombinant Protein (Q01) | |
VLDQLSAGKHLQSLVARGVGGSLGGGAGDDAEPLSKRHSDGIFTDSYSRYRKQMAVKKYLAAVLGKRYKQRVKNKGRR | |
RUO | |
ADCYAP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |