missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human B3GAT1 Partial ORF (ABM86204.1, 28 a.a. - 334 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | ABM86204.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 27087 |
Molecular Weight (g/mol) | 59.4kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16035415
|
Abnova™
H00027087-Q02.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16045415
|
Abnova™
H00027087-Q02.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the glucuronyltransferase gene family. These enzymes exhibit strict acceptor specificity, recognizing nonreducing terminal sugars and their anomeric linkages. This gene product functions as the key enzyme in a glucuronyl transfer reaction during the biosynthesis of the carbohydrate epitope HNK-1 (human natural killer-1, also known as CD57 and LEU7). Alternate transcriptional splice variants have been characterized. [provided by RefSeq]
Sequence: TLAPLLAVHKDEGSDPRRETPPGADPREYCTSDRDIVEVVRTEYVYTRPPPWSDTLPTIHVVTPTYSRPVQKAELTRMANTLLHVPNLHWLVVEDAPRRTPLTARLLRDTGLNYTHLHVETPRNYKLRGDARDPRIPRGTMQRNLALRWLRETFPRNSSQPGVVYFADDDNTYSLELFEEMRSTRRVSVWPVAFVGGLRYEAPRVNGAGKVVGWKTVFDPHRPFAIDMAGFAVNLRLILQRSQAYFKLRGVKGGYQESSLLRELVTLNDLEPKAANCTKILVWHTRTEKPVLVNEGKKGFTDPSVEISpecifications
ABM86204.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
59.4kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD57/GLCATP/GlcAT-P/GlcUAT-P/HNK-1/HNK1/LEU7/NK-1 | |
B3GAT1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
27087 | |
B3GAT1 (Human) Recombinant Protein (Q02) | |
TLAPLLAVHKDEGSDPRRETPPGADPREYCTSDRDIVEVVRTEYVYTRPPPWSDTLPTIHVVTPTYSRPVQKAELTRMANTLLHVPNLHWLVVEDAPRRTPLTARLLRDTGLNYTHLHVETPRNYKLRGDARDPRIPRGTMQRNLALRWLRETFPRNSSQPGVVYFADDDNTYSLELFEEMRSTRRVSVWPVAFVGGLRYEAPRVNGAGKVVGWKTVFDPHRPFAIDMAGFAVNLRLILQRSQAYFKLRGVKGGYQESSLLRELVTLNDLEPKAANCTKILVWHTRTEKPVLVNEGKKGFTDPSVEI | |
RUO | |
B3GAT1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |