missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human B7-2/CD86 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB141T-025
This item is not returnable.
View return policy
Description
B7-2/CD86 Monoclonal specifically detects B7-2/CD86 in Human samples. It is validated for Flow Cytometry.
Specifications
| B7-2/CD86 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD86 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human B7-2/CD86 | |
| 25 Tests | |
| Primary | |
| Detects human B7-2/CD86 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) B7-1, recombinant mouse B7-2, recombinant rat B7-2, rhB7-H1, rhB7-H2, rhB7-H3, rhB7-H3b, rhB7-H4, or rhB7-L2 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
| Flow Cytometry | |
| 37301 | |
| Flow Cytometry 5 uL/10^6 cells | |
| Activation B7-2 antigen, B70, B72, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86, CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 942 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction