missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BMP-4 Human ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EHBMP4

Additional Details : Weight : 0.00400kg

 View more versions of this product

Product Code. 15464913

  • 670.00 € / 1 set
Estimated Shipment: 23-04-2024
to see stock.
Add to basket

Description

Description

The Human Bone Morphogenic Protein 4 (Hu BMP-4) ELISA quantitates Hu BMP-4 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu BMP-4. Principle of the method The Human BMP-4 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The protein encoded by this gene is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. This particular family member plays an important role in the onset of endochondral bone formation in humans, and a reduction in expression has been associated with a variety of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. Alternative splicing in the 5′ untranslated region of this gene has been described and three variants are described, all encoding an identical protein.
Specifications

Specifications

P12644
15 pg/mL
Solid-phase sandwich ELISA
ELISA Kit
Plasma, Serum, Supernatant
96 assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
652
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Angiogenesis
2°C to 8°C
Human
4 hr. 45 min.
8.23-6000 pg/mL
15pg/mL to 6000pg/mL
Biotin
Growth Factors & Receptors
This ELISA kit shows no cross-reactivity with any of the cytokines tested: Human Angiogenin, BDNF, BLC, BMP-6, BMP-7, ENA-78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-7, IL-8, IL-9, IL- 10, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GMCSF, IFN-gamma, Leptin, MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF.
Human
Colorimetric Microplate Reader
BMP2B,BMP2B1,MCOPS6,OFC11,ZYME
<10%
HRP
RUO
Plasma, 50 μL; Serum, 50 μL; Supernatant, 100 μL
BMP2B, BMP2B1, MCOPS6, OFC11, ZYME
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers