missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CD25/IL-2R alpha Quantikine ELISA Kit
Human CD25/IL-2 R alpha Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Assay Range | 78.00 - 5000pg/mL |
---|---|
Assay Sensitivity | 10pg/mL |
Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum |
Description
The Human CD25/IL-2 R alpha Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human CD25/IL-2R alpha in cell culture supernates (10 ul), serum (13 ul), edta plasma (13 ul), heparin plasma (13 ul).Specifications
78.00 - 5000pg/mL | |
10pg/mL | |
Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum |