Learn More
Abnova™ Human CDKL3 Partial ORF (AAH41799.1, 322 a.a. - 420 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | AAH41799.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 51265 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16055505
|
Abnova™
H00051265-Q02.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16045505
|
Abnova™
H00051265-Q02.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This gene was identified as a gene absent in leukemic patients with chromosome 5q deletion. This loss may be an important determinant of dysmyelopoiesis. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: NELRKDERKTVYTNTLLSSSVLGKEIEKEKKPKEIKVRVIKVKGGRGDISEPKKKEYEGGLGQQDANENVHPMSPDTKLVTIEPPNPINPSTNCNGLKESpecifications
AAH41799.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NKIAMRE | |
CDKL3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
51265 | |
CDKL3 (Human) Recombinant Protein (Q02) | |
NELRKDERKTVYTNTLLSSSVLGKEIEKEKKPKEIKVRVIKVKGGRGDISEPKKKEYEGGLGQQDANENVHPMSPDTKLVTIEPPNPINPSTNCNGLKE | |
RUO | |
CDKL3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |