missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN-beta Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB814-500
1640.50 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IFN-beta Monoclonal specifically detects IFN-beta in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| IFN-beta | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3456 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Flow Cytometry, Neutralization | |
| 76703 | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 uL/10^6 cells, Neutralization 7-21 ug/mL | |
| P01574 | |
| IFNB1 | |
| E. coli-derived recombinant human IFN-beta Met22-Asn187 Accession # P01574 | |
| 500 μg | |
| Primary | |
| Detects human IFN-beta in Western blots. In Western blots, this antibody does not cross-react with recombinant human IFN-alpha . | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction