missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN-beta Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
213.00 € - 2050.00 €
Specifications
| Antigen | IFN-beta |
|---|---|
| Clone | 76703 |
| Dilution | Western Blot 1 ug/mL, Flow Cytometry 0.25 uL/10^6 cells, Neutralization 7-21 ug/mL |
| Applications | Western Blot, Flow Cytometry, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18786488
![]() |
R&D Systems
MAB814-SP |
25 μg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18706489
![]() |
R&D Systems
MAB814-100 |
100 μg |
519.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18752927
![]() |
R&D Systems
MAB814-500 |
500 μg |
2050.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN-beta Monoclonal specifically detects IFN-beta in Human samples. It is validated for Western Blot, Neutralization.Specifications
| IFN-beta | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 uL/10^6 cells, Neutralization 7-21 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P01574 | |
| 3456 | |
| E. coli-derived recombinant human IFN-beta Met22-Asn187 Accession # P01574 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IFN-beta in Western blots. In Western blots, this antibody does not cross-react with recombinant human IFN-alpha . |
| 76703 | |
| Western Blot, Flow Cytometry, Neutralization | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956 | |
| IFNB1 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title