missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse CD44 Rat, PerCP-Cy5.5, Clone: IM7, enQuireBio™
Rat Monoclonal Antibody
131.00 € - 276.00 €
Specifications
Antigen | Human/Mouse CD44 |
---|---|
Clone | IM7 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15991928
|
EnquireBio
QAB39-PCP55-25UG |
25 μg |
131.00 €
25µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15981928
|
EnquireBio
QAB39-PCP55-100UG |
100 μg |
276.00 €
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Human/Mouse CD44 | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1, pH7.2 | |
12505 | |
IgG2b κ | |
Protein A/G Purified |
IM7 | |
Flow Cytometry | |
PerCP-Cyanine5.5 | |
RUO | |
NP_001034239 | |
Cd44 | |
Primary | |
Store at 2-4°C. |