missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse CD45R / B220 Rat, APC-Cy7, Clone: RA3-6B2, enQuireBio™

Rat Monoclonal Antibody

Brand:  EnquireBio QAB41-APC7-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15991978

  • 287.45 € / 100µg

Estimated Shipment: 03-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Human/Mouse CD45R / B220
RA3-6B2
APC-Cyanine7
NP_001255215
Rat
Protein A/G Purified
RUO
Primary
19264
Flow Cytometry
0.2mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Ptprc
IgG2a kappa
100 μg
Store at 2-4°C.
Monoclonal
Videos
SDS
Documents
Certificates