missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PCTK2 Partial ORF (NP_002586, 426 a.a. - 523 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_002586 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 5128 |
Molecular Weight (g/mol) | 36.52kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16024515
|
Abnova™
H00005128-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16034515
|
Abnova™
H00005128-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene belongs to the cdc2/cdkx subfamily of the ser/thr family of protein kinases. It has similarity to rat protein which is thought to play a role in terminally differentiated neurons. [provided by RefSeq]
Sequence: NFPKYKPQPLINHAPRLDSEGIELITKFLQYESKKRVSAEEAMKHVYFRSLGPRIHALPESVSIFSLKEIQLQKDPGFRNSSYPETGHGKNRRQSMLFSpecifications
NP_002586 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PCTAIRE2 | |
PCTK2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
5128 | |
PCTK2 (Human) Recombinant Protein (Q01) | |
NFPKYKPQPLINHAPRLDSEGIELITKFLQYESKKRVSAEEAMKHVYFRSLGPRIHALPESVSIFSLKEIQLQKDPGFRNSSYPETGHGKNRRQSMLF | |
RUO | |
PCTK2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |